L(+)-Potassium sodium tartrate tetrahydrate, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ200

SIALS6170-1KGEA 237 EUR
SIALS6170-1KG SIALS6170-500G
L(+)-Potassium sodium tartrate tetrahydrate, Sigma-Aldrich®
L(+)-Potassium sodium tartrate tetrahydrate
Formula: KOOC(CHOH)₂COONa·4H₂O
MW: 282,22 g/mol
Melting Pt: 70…80 °C
Density: ∼1,767 g/cm³ (20 °C)
MDL Number: MFCD00150989
CAS Number: 6381-59-5
Merck Index: 13,07755
https://si.vwr.com/stibo/web/std.lang.all/98/18/2299818.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR