(S)-(+)-1-Aminopropan-2-ol, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: (S)-1-Aminopropan-2-ol, (S)-(+)-Isopropanolamine, (S)-(+)-1-Amino-2-propanol, (+)-1-Aminopropan-2-ol, (S)-Isopropanolamine, (+)-Isopropanolamine
MQ100
238864-1G 238864-5G
SIAL238864-1GEA 27.2 EUR
SIAL238864-1G SIAL238864-5G
(S)-(+)-1-Aminopropan-2-ol, Sigma-Aldrich®
(S)-(+)-1-Aminopropan-2-ol
Formula: C₃H₉NO
MW: 75.11 g/mol
Boiling Pt: 160
Melting Pt: –2 °C
Density: 0.96 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064429
CAS Number: 2799-17-9
EINECS: 220-533-4
UN: 3259
ADR: 8 II