(S)-(+)-2-Amino-1-butanol, Sigma-Aldrich®

Supplier: Merck
Danger

Synonyms: D(+)-2-Amino-1-butanol, (S)-2-Aminobutan-1-ol

M-Clarity™ quality level = MQ100

132527-1G 132527-5G
SIAL132527-1GEA 36.1 EUR
SIAL132527-1G SIAL132527-5G
(S)-(+)-2-Amino-1-butanol, Sigma-Aldrich®
(S)-(+)-2-Amino-1-butanol
Formula: C₄H₁₁NO
MW: 89.14 g/mol
Boiling Pt: 172 °C (1013 hPa)
Melting Pt: –2 °C
Density: 0.947 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064418
CAS Number: 5856-62-2
EINECS: 227-475-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR