(S)-(+)-2-Butanol, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: (S)-(+)-sec-Butyl alcohol, (S)-Butan-2-ol, (S)-(+)-sec-Butanol, (S)-(+)-Ethyl methyl carbinol, (S)-(+)-2-Hydroxy butane, (S)-(+)-Methyl ethyl carbinol
M-Clarity™ quality level = MQ100
237698-10G 237698-1G
SIAL237698-10GEA 424 EUR
SIAL237698-10G SIAL237698-1G
(S)-(+)-2-Butanol, Sigma-Aldrich®
(S)-(+)-2-Butanol
Formula: C₄H₁₀O
MW: 74.12 g/mol
Boiling Pt: 99 °C (1013 hPa)
Melting Pt: –114 °C
Density: 0.808 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064281
CAS Number: 4221-99-2
EINECS: 224-168-1
UN: 1120
ADR: 3,III
Merck Index: 13,01540