D-(-)-Sorbitol, Sigma-Aldrich®
Supplier: Merck
Synonyms: (2R,3R,4R,5S)-Hexane-1,2,3,4,5,6-hexaol, (Sorbitol), D(-)-Glucitol, D-Sorbitol, Sorbitol
MQ300
S1876-500G S1876-100G S1876-10KG S1876-10MG S1876-1KG S1876-5KG
SIALS1876-500GEA 41.5 EUR
SIALS1876-500G SIALS1876-100G SIALS1876-10KG SIALS1876-10MG SIALS1876-1KG SIALS1876-5KG
D-(-)-Sorbitol, Sigma-Aldrich®
D(-)-Sorbitol
Formula: HOH₂C(CH(OH))₄CH₂OH
MW: 182.17 g/mol
Boiling Pt: 295
Melting Pt: 94…98 °C
Density: 1.524 g/cm³ (20 °C)
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00004708
CAS Number: 50-70-4
EINECS: 200-061-5
Merck Index: 13,08797
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f82054e8cfed2cf3e8b739c3cb816c4f91bd2735&vid=a084778ac67fbdf7e15478c0db755e78cf3d95c9&prid=web&clid=SAPDAM