(S)-(+)-1-Bromo-2-methylbutane, Sigma-Aldrich®

Supplier: Merck
Danger

Synonyms: (S)-(+)-2-Methylbutyl bromide

M-Clarity™ quality level = MQ100

250023-1G 250023-5G
SIAL250023-1GEA 85.4 EUR
SIAL250023-1G SIAL250023-5G
(S)-(+)-1-Bromo-2-methylbutane, Sigma-Aldrich®
(S)-(+)-1-Bromo-2-methylbutane
Formula: C₅H₁₁Br
MW: 151.05 g/mol
Boiling Pt: 122…123 °C (1013 hPa)
Density: 1.221 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00078662
CAS Number: 534-00-9
EINECS: 208-583-5
UN: 1993
ADR: 3,II
Merck Index: 12,00642

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR