ESD occupational shoes, clogs, C-FIT, Levy
Supplier: ELTEN
972460/44 972460/45 972460/46 972460/47 972460/48 972460/40 972460/41
ELTN972460/44EA 53.9 EUR
ELTN972460/44 ELTN972460/45 ELTN972460/46 ELTN972460/47 ELTN972460/48 ELTN972460/40 ELTN972460/41
ESD occupational shoes, clogs, C-FIT, Levy
Shoes and Boots
Leather-free, lightweight clogs made from smooth, tear resistant microfibre upper, an adjustable and foldable heel strap, plus a mono-density PU sole (2,5 mm, oil, fuel and abrasion resistant, also heat resistant up to approx. 130 °C).
  • Climate-regulating, breathable textile lining
  • Full length inlay sole (C-Fit ESD)
  • ESD soft fleece insole
  • Anatomically-shaped insole and ankle support for improved stability
  • Shock-absorbent heel
Colour: White
Certifikace: EN 61340-5-1 (ESD); EN ISO 20347; OB, SRC, A, E, FO