L-Norvaline, Sigma-Aldrich®
Supplier: Merck
Synonyms: (S)-(+)-Norvaline, L(+)-Norvaline, H-Nva-OH, (S)-2-Aminopentanoic acid
M-Clarity™ quality level = MQ200
N7627-10G N7627-1G N7627-5G
SIALN7627-10GEA 348 EUR
SIALN7627-10G SIALN7627-1G SIALN7627-5G
L-Norvaline, Sigma-Aldrich®
L-Norvaline
Formula: C₅H₁₁NO₂
MW: 117.15 g/mol
Boiling Pt: 228 °C (1013 hPa)
Melting Pt: 101 °C
MDL Number: MFCD00064421
CAS Number: 6600-40-4
EINECS: 229-543-3
Merck Index: 12,06813