D-(-)-Histidine, Sigma-Aldrich®

Supplier: Merck

Synonyms: (R)-(-)-Histidine, D-3-Imidazol-4-ylalanine, (2R)-(-)-Histidine, H-D-His-OH

M-Clarity™ quality level = MQ200

H3751-25G H3751-5G
SIALH3751-25GEA 663 EUR
SIALH3751-25G SIALH3751-5G
D-(-)-Histidine, Sigma-Aldrich®
D(-)-Histidine
Formula: C₆H₉N₃O₂
MW: 155.16 g/mol
Melting Pt: 287 °C
MDL Number: MFCD00065963
CAS Number: 351-50-8
EINECS: 206-513-8
Merck Index: 12,04758

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR