L(-)-Tryptophan, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ200
T0254-25G T0254-100G T0254-1G T0254-1KG T0254-500G T0254-5G
SIALT0254-25GEA 67.1 EUR
SIALT0254-25G SIALT0254-100G SIALT0254-1G SIALT0254-1KG SIALT0254-500G SIALT0254-5G
L(-)-Tryptophan, Sigma-Aldrich®
L(-)-Tryptophan
Formula: C₁₁H₁₂N₂O₂
Melting Pt: 290 °C
Density: 1.34 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064340
CAS Number: 73-22-3
EINECS: 200-795-6
Merck Index: 12,09929