L(-)-Tyrosine, Sigma-Aldrich®

Supplier: Merck

Synonyms: L-Tyrosine, (S)-(-)-Tyrosine, Tyrosine, H-Tyr-OH

M-Clarity™ quality level = MQ200

T8566-100G T8566-10MG T8566-1KG T8566-25G
SIALT8566-100GEA 257 EUR
SIALT8566-100G SIALT8566-10MG SIALT8566-1KG SIALT8566-25G
L(-)-Tyrosine, Sigma-Aldrich®
L(-)-Tyrosine
Formula: C₉H₁₁NO₃
MW: 181.19 g/mol
Boiling Pt: 351 °C (1013 hPa)
Melting Pt: 285 °C
Density: 1.41 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002606
CAS Number: 60-18-4
EINECS: 200-460-4
Merck Index: 13,09907

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR