D-(+)-Camphor, Sigma-Aldrich®

Supplier: Merck
Warning

Synonyms: (R)-(+)-1,7,7-Trimethylbicyclo[2.2.1]-2-heptanone, (1R)-(+)-1,7,7-Trimethyl-2-norbornanone, (1R,4R)-1,7,7-Trimethylbicyclo[2.2.1]heptan-2-one, (1R,4R)-(+)-1,7,7-Trimethylbicyclo[2.2.1]-2-heptanone, (1R,4R)-(+)-1,7,7-Trimethyl-2-norbornanone, (R)-(+)-1,7,7-Trimethyl-2-norbornanone, (1R)-(+)-1,7,7-Trimethylbicyclo[2.2.1]-2-heptanone, (+)-Camphor

M-Clarity™ quality level = MQ300

W223018-10KG-K W223018-1KG-K
SIALW223018-10KG-KEA 494 EUR
SIALW223018-10KG-K SIALW223018-1KG-K
D-(+)-Camphor, Sigma-Aldrich®
D(+)-Camphor
Formula: C₁₀H₁₆O
MW: 152.24 g/mol
Boiling Pt: 408 °C (1013 hPa)
Melting Pt: 174…175 °C
Density: 0.992 g/cm³ (20 °C)
Flash Pt: 66 °C
Storage Temperature: Ambient
MDL Number: MFCD00064149
CAS Number: 464-49-3
EINECS: 207-355-2
UN: 2717
ADR: 4.1,III
Merck Index: 13,01739

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR