(R)-(+)-Limonene, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: (R)-(+)-Dipentene, (R)-(+)-1-Methyl-4-(1-methylvinyl)cyclohexene, (R)-(+)-4-Isopropenyl-1-methyl-1-cyclohexene, D(+)-Limonene, (R)-(+)-p-Mentha-1,8-diene, D-Limonene
M-Clarity™ quality level = MQ300
W263303-1KG-K W263303-20KG-K
SIALW263303-1KG-KEA 84.4 EUR
SIALW263303-1KG-K SIALW263303-20KG-K
(R)-(+)-Limonene, Sigma-Aldrich®
(R)-(+)-Limonene
Formula: C₁₀H₁₆
MW: 136.24 g/mol
Boiling Pt: 175.5 °C (1013 hPa)
Melting Pt: –74 °C
Density: 0.8411 g/cm³ (20 °C)
Flash Pt: 43 °C
Storage Temperature: Ambient
MDL Number: MFCD00062991
CAS Number: 5989-27-5
EINECS: 227-813-5
UN: 2052
ADR: 3,III
Merck Index: 13,05515