1,10-Phenanthroline, Sigma-Aldrich®
Supplier: Merck
|
Danger
|
Synonyms:
o-Phenanthroline
M-Clarity™ quality level = MQ200
131377-100G
131377-2.5G
131377-25G
131377-5G
SIAL131377-100GEA
249
EUR
SIAL131377-100G
SIAL131377-2.5G
SIAL131377-25G
SIAL131377-5G
1,10-Phenanthroline, Sigma-Aldrich®
1,10-Phenanthroline
Formula:
C₁₂H₈N₂ MW: 180.21 g/mol Boiling Pt: 360 °C (1013 hPa) Melting Pt: 117…119 °C |
MDL Number:
MFCD00011678 CAS Number: 66-71-7 EINECS: 200-629-2 UN: 2811 ADR: 6.1,III Merck Index: 12,07356 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...