1,10-Phenanthroline monohydrate, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: 1,10-Phenanthroline hydrate
M-Clarity™ quality level = MQ200
P9375-100G P9375-1G P9375-25G P9375-5G
SIALP9375-100GEA 828 EUR
SIALP9375-100G SIALP9375-1G SIALP9375-25G SIALP9375-5G
1,10-Phenanthroline monohydrate, Sigma-Aldrich®
1,10-Phenanthroline monohydrate
Formula: C₁₂H₈N₂·1H₂O
MW: 198.22 g/mol
Melting Pt: 93…94 °C
Density: 1.259 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149973
CAS Number: 5144-89-8
EINECS: 200-629-2
UN: 2811
ADR: 6.1 III
Merck Index: 13,07291
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=aba346b9f67fdda9afd4d48b893e6318564d604c&vid=2d08e04ee9525293a747b55cdcff8f66a6c4715a&prid=web&clid=SAPDAM