Ethyl linoleate, Sigma-Aldrich®

Supplier: Merck

Synonyms: 9Z,12Z-Octadecadienoic acid ethyl ester, Linoleic acid ethyl ester, cis,cis-9,12-Octadecadienoic acid ethyl ester, (9Z,12Z)-Ethyl octadeca-9,12-dienoate

M-Clarity™ quality level = MQ200

L1751-5G L1751-1G
SIALL1751-5GEA 179 EUR
SIALL1751-5G SIALL1751-1G
Ethyl linoleate, Sigma-Aldrich®
Ethyl linoleate
Formula: C₂₀H₃₆O₂
MW: 308.5 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00009535
CAS Number: 544-35-4
EINECS: 208-868-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR