Methyl linoleate

Supplier: Roth Carl

Synonyms: cis,cis-9,12-Octadecadienoic acid methyl ester, 9,12-Octadecanoic acid methyl ester, 9Z,12Z-Octadecadienoic acid methyl ester, Linoleic acid methyl ester, (9Z,12Z)-Methyl octadeca-9,12-dienoate, Methyl cis,cis-9,12-octadecadienoate, 9Z,12Z-Oktadekadiensäuremethylester, 9,12 Methyl octadecaotenoate methyl linoleate, cis,cis-9,12-Oktadekadiensäuremethylester

7217.1 7217.2
ROTH7217.1EA 63.8 EUR
ROTH7217.1 ROTH7217.2
Methyl linoleate
Methyl linoleate
Formula: C₁₉H₃₄O₂
MW: 294.48 g/mol
Boiling Pt: 340 °C (1013 hPa)
Melting Pt: –35 °C
Density: 0,89 g/cm³ (20 °C)
MDL Number: MFCD00009534
CAS Number: 112-63-0
EINECS: 203-993-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR