Methyl linoleate, Sigma-Aldrich®

Supplier: Merck

Synonyms: cis,cis-9,12-Octadecadienoic acid methyl ester, 9,12-Octadecanoic acid methyl ester, 9Z,12Z-Octadecadienoic acid methyl ester, Linoleic acid methyl ester, (9Z,12Z)-Methyl octadeca-9,12-dienoate, Methyl cis,cis-9,12-octadecadienoate, 9Z,12Z-Oktadekadiensäuremethylester, 9,12 Methyl octadecaotenoate methyl linoleate, cis,cis-9,12-Oktadekadiensäuremethylester

M-Clarity™ quality level = MQ200

L1876-5G L1876-25G L1876-10G
SIALL1876-5GEA 255 EUR
SIALL1876-5G SIALL1876-25G SIALL1876-10G
Methyl linoleate, Sigma-Aldrich®
Methyl linoleate
Formula: C₁₉H₃₄O₂
MW: 294.48 g/mol
Boiling Pt: 340
Melting Pt: –35 °C
Density: 0.89 g/cm³ (20 °C)
MDL Number: MFCD00009534
CAS Number: 112-63-0
EINECS: 203-993-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR