Linoleic acid
Supplier: Cayman Chemical
Synonyms: (9Z,12Z)-Octadeca-9,12-dienoic acid, cis,cis-9,12-Octadecadienoic acid, 9Z,12Z-Octadecadienoic acid
90150-100 90150-50 90150-500
CAYM90150-100EA 39 EUR
CAYM90150-100 CAYM90150-50 CAYM90150-500
Linoleic acid
Linoleic acid
Formula: C₁₈H₃₂O₂
MW: 280.45 g/mol
Boiling Pt: 360 °C (1013 hPa)
Melting Pt: –5 °C
Density: 0.9 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064241
CAS Number: 60-33-3
EINECS: 200-470-9
Merck Index: 13,05527