Linoleic acid, Sigma-Aldrich®
Supplier: Merck
Synonyms: (9Z,12Z)-Octadeca-9,12-dienoic acid, cis,cis-9,12-Octadecadienoic acid, 9Z,12Z-Octadecadienoic acid
MQ200
L1012-100MG L1012-1G L1012-5G
SIALL1012-100MGEA 37.3 EUR
SIALL1012-100MG SIALL1012-1G SIALL1012-5G
Linoleic acid, Sigma-Aldrich®
Linoleic acid
Formula: C₁₈H₃₂O₂
MW: 280.45 g/mol
Boiling Pt: 360
Melting Pt: –5 °C
Density: 0.9 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064241
CAS Number: 60-33-3
EINECS: 200-470-9
Merck Index: 13,05527