Linoleic acid, Sigma-Aldrich®
Supplier: Merck
Synonyms:
(9Z,12Z)-Octadeca-9,12-dienoic acid, cis,cis-9,12-Octadecadienoic acid, 9Z,12Z-Octadecadienoic acid
MQ200
L1376-5G
L1376-10G
L1376-10MG
L1376-1G
L1376-25G
L1376-500MG
SIALL1376-5GEA
111
EUR
SIALL1376-5G
SIALL1376-10G
SIALL1376-10MG
SIALL1376-1G
SIALL1376-25G
SIALL1376-500MG
Linoleic acid, Sigma-Aldrich®
Linoleic acid
Formula:
C₁₈H₃₂O₂ MW: 280.45 g/mol Boiling Pt: 360 Melting Pt: –5 °C Density: 0.9 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00064241 CAS Number: 60-33-3 EINECS: 200-470-9 Merck Index: 13,05527 |