1,4-Pentadiene, Sigma-Aldrich®
Supplier: Merck
Synonyms: Penta-1,4-diene
M-Clarity™ quality level = MQ100
P4607-1G P4607-5G
SIALP4607-1GEA 57.5 EUR
SIALP4607-1G SIALP4607-5G
1,4-Pentadiene, Sigma-Aldrich®
1,4-Pentadiene
Formula: C₅H₈
Boiling Pt: 26 °C (1013 hPa)
Melting Pt: –148.3 °C
Density: 0.6608 g/cm³ (20 °C)
MDL Number: MFCD00008665
CAS Number: 591-93-5
EINECS: 209-736-9
UN: 3295
ADR: 3,I