1,5-Hexadiene, Sigma-Aldrich®
Supplier: Merck
Synonyms: Hexa-1,5-diene
M-Clarity™ quality level = MQ100
128554-100G 128554-25G
SIAL128554-100GEA 109 EUR
SIAL128554-100G SIAL128554-25G
1,5-Hexadiene, Sigma-Aldrich®
1,5-Hexadiene
Formula: C₆H₁₀
MW: 82.15 g/mol
Boiling Pt: 60 °C (1013 hPa)
Melting Pt: –141 °C
Density: 0.692 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008666
CAS Number: 592-42-7
EINECS: 209-754-7
UN: 2458
ADR: 3,II