1,8-Nonadiyne, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ100

SIAL161306-10GEA 160 EUR
SIAL161306-10G
1,8-Nonadiyne, Sigma-Aldrich®
1,8-Nonadiyne
Formula: C₉H₁₂
MW: 120,19 g/mol
Boiling Pt: 162 °C (1013 hPa)
Melting Pt: –27,3 °C
Density: 0,8158 g/cm³ (20 °C)
MDL Number: MFCD00008581
UN: 3295
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR