1,1,2,2-Tetrabromoethane, Sigma-Aldrich®
Supplier: Merck
Danger
M-Clarity™ quality level = MQ100
185574-1KG 185574-250G
SIAL185574-1KGEA 111 EUR
SIAL185574-1KG SIAL185574-250G
1,1,2,2-Tetrabromoethane, Sigma-Aldrich®
1,1,2,2-Tetrabromoethane
Formula: Br₂HCCHBr₂
MW: 345.65 g/mol
Boiling Pt: 244 °C (1013 hPa)
Melting Pt: –1 °C
Density: 2.9656 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000133
CAS Number: 79-27-6
EINECS: 201-191-5
UN: 2504
ADR: 6.1,III
Merck Index: 13,09261
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ecc5c2f2399032fc65a1f90353ec556de6f2f655&vid=602784cfe0c9c19c147c1108e11d03d302f55362&prid=web&clid=SAPDAM