1,4-Dibromobutane, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: 1,4-Dibromo butane
M-Clarity™ quality level = MQ100
140805-100G 140805-500G
SIAL140805-100GEA 43.5 EUR
SIAL140805-100G SIAL140805-500G
1,4-Dibromobutane, Sigma-Aldrich®
1,4-Dibromobutane
Formula: BrH₂C(CH₂)₂CH₂Br
MW: 215.92 g/mol
Boiling Pt: 197 °C (1013 hPa)
Melting Pt: –20 °C
Density: 1.83 g/cm³ (20 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00000261
CAS Number: 110-52-1
EINECS: 203-775-5
UN: 2810
ADR: 6.1,II