1,6-Dibromohexane, Sigma-Aldrich®
Supplier: Merck
Synonyms: Hexamethylen dibromide
M-Clarity™ quality level = MQ200
D41007-100G D41007-25G D41007-500G
SIALD41007-100GEA 37.2 EUR
SIALD41007-100G SIALD41007-25G SIALD41007-500G
1,6-Dibromohexane, Sigma-Aldrich®
1,6-Dibromohexane
Formula: BrH₂C(CH₂)₄CH₂Br
MW: 243.97 g/mol
Boiling Pt: 243 °C (1013 hPa)
Melting Pt: –2 °C
Density: 1.61 g/cm³ (20 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00000272
CAS Number: 629-03-8
EINECS: 211-067-2
UN: 3082
ADR: 9,III