1,7-Dibromoheptane, Sigma-Aldrich®
Supplier: Merck
Synonyms: Heptamethylene dibromide
M-Clarity™ quality level = MQ100
144991-10G 144991-50G
SIAL144991-10GEA 109 EUR
SIAL144991-10G SIAL144991-50G
1,7-Dibromoheptane, Sigma-Aldrich®
1,7-Dibromoheptane
Formula: BrH₂C(CH₂)₅CH₂Br
MW: 258 g/mol
Boiling Pt: 255 °C (1013 hPa)
Melting Pt: –41.7 °C
Density: 1.5451 g/cm³ (0 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000274
CAS Number: 4549-31-9
EINECS: 224-910-4
UN: 2810
ADR: 6.1,III