1-Bromobutane, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Butyl bromide
M-Clarity™ quality level = MQ200
B59497-1KG B59497-2.5KG B59497-25G B59497-500G
SIALB59497-1KGEA 86.8 EUR
SIALB59497-1KG SIALB59497-2.5KG SIALB59497-25G SIALB59497-500G
1-Bromobutane, Sigma-Aldrich®
1-Bromobutane
Formula: BrH₂C(CH₂)₂CH₃
Boiling Pt: 101 °C (1013 hPa)
Melting Pt: –112 °C
Density: 1.276 g/cm³ (20 °C)
Flash Pt: 16 °C
Storage Temperature: Ambient
MDL Number: MFCD00000260
CAS Number: 109-65-9
EINECS: 203-691-9
UN: 1126
ADR: 3,II
Merck Index: 13,01552