1-Bromohexane, Sigma-Aldrich®
Supplier: Merck
Synonyms: 1-Hexyl bromide
M-Clarity™ quality level = MQ200
B68240-100G B68240-1KG B68240-500G
SIALB68240-100GEA 22.2 EUR
SIALB68240-100G SIALB68240-1KG SIALB68240-500G
1-Bromohexane, Sigma-Aldrich®
1-Bromohexane
Formula: BrH₂C(CH₂)₄CH₃
MW: 165.07 g/mol
Boiling Pt: 155 °C (1013 hPa)
Melting Pt: –85 °C
Density: 1.167 g/cm³ (20 °C)
Flash Pt: 47 °C
Storage Temperature: Ambient
MDL Number: MFCD00000271
CAS Number: 111-25-1
EINECS: 203-850-2
UN: 1993
ADR: 3,III