1-Bromooctadecane, Sigma-Aldrich®

Supplier: Merck

199494-100G 199494-500G
SIAL199494-100GEA 29.5 EUR
SIAL199494-100G SIAL199494-500G
1-Bromooctadecane, Sigma-Aldrich®
1-Bromooctadecane
Formula: BrH₂C(CH₂)₁₆CH₃
MDL Number: MFCD00000231
CAS Number: 112-89-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR