1-Bromooctane, Sigma-Aldrich®
Supplier: Merck
|
Warning
|
Synonyms:
1-Octyl bromide
M-Clarity™ quality level = MQ200
152951-100G
152951-500G
SIAL152951-100GEA
24.9
EUR
SIAL152951-100G
SIAL152951-500G
1-Bromooctane, Sigma-Aldrich®
1-Bromooctane
Formula:
BrH₂C(CH₂)₆CH₃ MW: 193.13 g/mol Boiling Pt: 198…201 °C (1013 hPa) Melting Pt: –55 °C Density: 1.112 g/cm³ (20 °C) Storage Temperature: Freezer |
MDL Number:
MFCD00000276 CAS Number: 111-83-1 EINECS: 203-912-9 UN: 3082 ADR: 9,III Merck Index: 12,06861 |
Learn more
![](/m-si.vwr.com/sl_SI/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...