1-Bromotetradecane, Sigma-Aldrich®

Supplier: Merck

Synonyms: Myristyl bromide

M-Clarity™ quality level = MQ200

195332-500ML 195332-5ML
SIAL195332-500MLEA 94.7 EUR
SIAL195332-500ML SIAL195332-5ML
1-Bromotetradecane, Sigma-Aldrich®
1-Bromotetradecane
Formula: BrH₂C(CH₂)₁₂CH₃
MW: 277.29 g/mol
Boiling Pt: 308 °C (1013 hPa)
Melting Pt: 5 °C
Density: 1.02 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000228
CAS Number: 112-71-0
EINECS: 203-999-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR