Bromoacetonitrile, Sigma-Aldrich®
Supplier: Merck
Danger
M-Clarity™ quality level = MQ200
242489-100G 242489-25G 242489-5G
SIAL242489-100GEA 263 EUR
SIAL242489-100G SIAL242489-25G SIAL242489-5G
Bromoacetonitrile, Sigma-Aldrich®
Bromoacetonitrile
Formula: BrCNCH₂
Boiling Pt: 150 °C (1013 hPa)
Melting Pt: –36 °C
Density: 1.771 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00001884
CAS Number: 590-17-0
EINECS: 209-672-1
UN: 3276
ADR: 6.1,II