Ethyl bromopyruvate, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: Bromopyruvic acid ethyl ester, Ethyl 3-bromo-2-oxopropanoate, 3-Bromo-2-oxopropionic acid ethyl ester
M-Clarity™ quality level = MQ100
E14578-100G E14578-25G E14578-5G
SIALE14578-100GEA 266 EUR
SIALE14578-100G SIALE14578-25G SIALE14578-5G
Ethyl bromopyruvate, Sigma-Aldrich®
Ethyl bromopyruvate
Formula: BrH₂CC(O)COOCH₂CH₃
MW: 195.01 g/mol
Boiling Pt: 98…100 °C (13 hPa)
Density: 1.55 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000204
CAS Number: 70-23-5
EINECS: 200-729-6
UN: 3265
ADR: 8 II