4'-Bromoacetanilide, Sigma-Aldrich®

Supplier: Merck

Synonyms: 4-Bromoacetanilide, N-(4-Bromophenyl)acetamide

M-Clarity™ quality level = MQ100

161659-100G 161659-25G
SIAL161659-100GEA 263 EUR
SIAL161659-100G SIAL161659-25G
4'-Bromoacetanilide, Sigma-Aldrich®
4'-Bromoacetanilide
Formula: H₃CC(O)NH(C₆H₄Br)
MW: 214.06 g/mol
Boiling Pt: 347 °C (1013 hPa)
Melting Pt: 164…167 °C
Density: 1.717 g/cm³ (20 °C)
MDL Number: MFCD00000092
CAS Number: 103-88-8
EINECS: 203-154-9
Merck Index: 13,01383

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR