4-Bromobutyric acid, Sigma-Aldrich®
Supplier: Merck
|
Danger
|
Synonyms:
4-Bromobutanoic acid
M-Clarity™ quality level = MQ100
258083-10G
258083-50G
SIAL258083-10GEA
46.4
EUR
SIAL258083-10G
SIAL258083-50G
4-Bromobutyric acid, Sigma-Aldrich®
4-Bromobutyric acid
Formula:
BrH₂C(CH₂)₂COOH Boiling Pt: 249 °C (1013 hPa) Melting Pt: 29 °C |
MDL Number:
MFCD00002817 CAS Number: 2623-87-2 EINECS: 220-083-9 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...