5-Bromovaleric acid, Sigma-Aldrich®

Supplier: Merck
Warning

Synonyms: 5-Bromopentanoic acid

MQ100

158410-10G 158410-50G
SIAL158410-10GEA 38.1 EUR
SIAL158410-10G SIAL158410-50G
5-Bromovaleric acid, Sigma-Aldrich®
5-Bromovaleric acid
Formula: BrH₂C(CH₂)₃COOH
MW: 181.03 g/mol
Boiling Pt: 266
Melting Pt: 38…40 °C
Storage Temperature: Ambient
MDL Number: MFCD00004414
CAS Number: 2067-33-6
EINECS: 218-185-3
UN: 3261
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR