6-Bromocaproic acid, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: 6-Bromohexanoic acid
M-Clarity™ quality level = MQ200
150452-100G 150452-25G 150452-5G
SIAL150452-100GEA 125 EUR
SIAL150452-100G SIAL150452-25G SIAL150452-5G
6-Bromocaproic acid, Sigma-Aldrich®
6-Bromocaproic acid
Formula: BrH₂C(CH₂)₄COOH
MW: 195.06 g/mol
Boiling Pt: 165 °C (1013 hPa)
Melting Pt: 31…34 °C
Density: 1.4068 g/cm³ (30 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004422
CAS Number: 4224-70-8
EINECS: 224-176-5
UN: 3261
ADR: 8,III