Bromoacetic acid ≥99.0%, ReagentPlus®, Sigma-Aldrich®
Supplier: Merck
|
Danger
|
M-Clarity™ quality level = MQ200
17000-100G
17000-25G
17000-500G
SIAL17000-100GEA
96.8
EUR
SIAL17000-100G
SIAL17000-25G
SIAL17000-500G
Bromoacetic acid ≥99.0%, ReagentPlus®, Sigma-Aldrich®
Bromoacetic acid
Formula:
BrCH₂COOH MW: 138.95 g/mol Boiling Pt: 206 °C (1013 hPa) Melting Pt: 46…49 °C Density: 1.9335 g/cm³ (20 °C) Flash Pt: >112 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002678 CAS Number: 79-08-3 EINECS: 201-175-8 UN: 3425 ADR: 8,II Merck Index: 13,01384 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...