1-Bromo-2-methoxyethane, Sigma-Aldrich®

Supplier: Merck

Synonyms: 2-Bromoethyl methyl ether

M-Clarity™ quality level = MQ100

238155-25G 238155-5G
SIAL238155-25GEA 73.8 EUR
SIAL238155-25G SIAL238155-5G
1-Bromo-2-methoxyethane, Sigma-Aldrich®
1-Bromo-2-methoxyethane
Formula: BrH₂CCH₂OCH₃
MW: 138.99 g/mol
Boiling Pt: 110
Density: 1.46 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00000236
CAS Number: 6482-24-2
EINECS: 229-339-4
UN: 3271
ADR: 3 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR