Bromoacetaldehyde dimethyl acetal, Sigma-Aldrich®
Supplier: Merck
Danger
M-Clarity™ quality level = MQ100
242500-100G 242500-25G
SIAL242500-100GEA 236 EUR
SIAL242500-100G SIAL242500-25G
Bromoacetaldehyde dimethyl acetal, Sigma-Aldrich®
Bromoacetaldehyde dimethyl acetal
Formula: BrH₂CCH(OCH₃)₂
Boiling Pt: 148 °C (1013 hPa)
Density: 1.43 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00000213
CAS Number: 7252-83-7
EINECS: 230-669-6
UN: 2929
ADR: 6.1,II