2,2,2-Tribromoethanol, Sigma-Aldrich®

Supplier: Merck
Warning

Synonyms: Tribromoethyl alcohol

M-Clarity™ quality level = MQ200

T48402-100G T48402-25G T48402-5G
SIALT48402-100GEA 316 EUR
SIALT48402-100G SIALT48402-25G SIALT48402-5G
2,2,2-Tribromoethanol, Sigma-Aldrich®
2,2,2-Tribromoethanol
Formula: Br₃CCH₂OH
MW: 282.76 g/mol
Boiling Pt: 92…93 °C (10 mmHg)
Melting Pt: 81 °C
Storage Temperature: Ambient
MDL Number: MFCD00004671
CAS Number: 75-80-9
EINECS: 200-903-1
Merck Index: 12,09743

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR