3-Bromo-1-propanol, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: 3-Bromopropan-1-ol, 1-Bromo-3-hydroxypropane
M-Clarity™ quality level = MQ100
167169-100G 167169-25G 167169-5G
SIAL167169-100GEA 387 EUR
SIAL167169-100G SIAL167169-25G SIAL167169-5G
3-Bromo-1-propanol, Sigma-Aldrich®
3-Bromo-1-propanol
Formula: BrH₂CCH₂CH₂OH
MW: 138.99 g/mol
Boiling Pt: 145 °C (1013 hPa)
Density: 1.6 g/cm³ (20 °C)
Flash Pt: 65 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00002942
CAS Number: 627-18-9
EINECS: 210-986-6