6-Bromo-1-hexanol 95%
Supplier: Thermo Fisher Scientific
Warning
Synonyms: 6-Bromohexan-1-ol, Hexamethylene bromohydrin, 1-Bromo-6-hydroxyhexane
270090025 270090100 270090500
ACRO270090025EA 68 EUR
ACRO270090025 ACRO270090100 ACRO270090500
6-Bromo-1-hexanol 95%
6-Bromo-1-hexanol
Formula: BrH₂C(CH₂)₄CH₂OH
MW: 181.07 g/mol
Boiling Pt: 104…106 °C (5 mmHg)
Density: 1.32 g/cm³
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00002983
CAS Number: 4286-55-9
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4810 to 1.4840 (20°C, 589 nm)
GC ≥94 %