Butyraldehyde, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Butylaldehyde, Butanal
M-Clarity™ quality level = MQ300
W221910-100G-K W221910-1KG-K W221910-25G-K
SIALW221910-100G-KEA 160 EUR
SIALW221910-100G-K SIALW221910-1KG-K SIALW221910-25G-K
Butyraldehyde, Sigma-Aldrich®
Butyraldehyde
Formula: C₄H₈O
MW: 72.11 g/mol
Boiling Pt: 76 °C (1013 hPa)
Melting Pt: –99 °C
Density: 0.803 g/cm³ (20 °C)
Flash Pt: –12 °C
Storage Temperature: Ambient
MDL Number: MFCD00007023
CAS Number: 123-72-8
EINECS: 204-646-6
UN: 1129
ADR: 3,II
Merck Index: 13,01591