Caprinaldehyde, Sigma-Aldrich®
Supplier: Merck
Synonyms: Decylaldehyde, Decanal, Decyl aldehyde
M-Clarity™ quality level = MQ300
W236209-1KG-K W236209-20KG-K W236209-8KG-K
SIALW236209-1KG-KEA 80.2 EUR
SIALW236209-1KG-K SIALW236209-20KG-K SIALW236209-8KG-K
Caprinaldehyde, Sigma-Aldrich®
Caprinaldehyde
Formula: C₁₀H₂₀O
Boiling Pt: 207 °C (1013 hPa)
Melting Pt: 7 °C
Density: 0.83 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00007031
CAS Number: 112-31-2
EINECS: 203-957-4