Laurinaldehyde, Sigma-Aldrich®

Supplier: Merck

Synonyms: Lauric aldehyde, Dodecylaldehyde, Dodecanal

M-Clarity™ quality level = MQ100

D222003-100G D222003-2G
SIALD222003-100GEA 89.5 EUR
SIALD222003-100G SIALD222003-2G
Laurinaldehyde, Sigma-Aldrich®
Laurinaldehyde
Formula: C₁₂H₂₄O
Boiling Pt: 242 °C (1013 hPa)
Melting Pt: 44 °C
Density: 0.83 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00007017
CAS Number: 112-54-9
EINECS: 203-983-6
UN: 3082
ADR: 9,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR