Laurinaldehyde, Sigma-Aldrich®

Supplier: Merck

Synonyms: Lauric aldehyde, Dodecylaldehyde, Dodecanal

M-Clarity™ quality level = MQ300

W261505-1KG-K W261505-20KG-K W261505-4KG-K
SIALW261505-1KG-KEA 117 EUR
SIALW261505-1KG-K SIALW261505-20KG-K SIALW261505-4KG-K
Laurinaldehyde, Sigma-Aldrich®
Laurinaldehyde
Formula: C₁₂H₂₄O
MW: 184.32 g/mol
Boiling Pt: 242
Melting Pt: 44
Density: 0.83 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00007017
CAS Number: 112-54-9
EINECS: 203-983-6
UN: 3082
ADR: 9 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR