Acetaldehyde, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Ethanal
M-Clarity™ quality level = MQ300
W200336-1.9KG- W200336-12KG-K W200336-400G-K
SIALW200336-1.9KG-EA 658 EUR
SIALW200336-1.9KG- SIALW200336-12KG-K SIALW200336-400G-K
Acetaldehyde, Sigma-Aldrich®
Acetaldehyde
Formula: CH₃CHO
MW: 44.05 g/mol
Boiling Pt: 20.1 °C (1013 hPa)
Melting Pt: –123 °C
Density: 0.78 g/cm³ (20 °C)
Flash Pt: –27 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00006991
CAS Number: 75-07-0
EINECS: 200-836-8
UN: 1089
ADR: 3,I
Merck Index: 12,00037
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=de03379d171eda3c733a039cd16293e8ee4a02e2&vid=573943501bd6753f30bb12e41521b99361988998&prid=web&clid=SAPDAM